Would you like to log in again?
TETPOR AIR Filter Cartridges, Parker Bioscience
Supplier: Parker
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
ZCMT3-020E-AE ZCMT3-020C-AE ZCMT2-020E-A ZCMT2-020C-A ZCMTK-020D-AE ZCMT3-020D-A ZCMT1-020D-AE ZCMTK-020D-A ZCMT1-020E-A ZCMT2-020D-AE ZCMTK-020E-AE ZCMT1-020C-A ZCMT2-020C-AE ZCMTK/020E-A ZCMT3-020C-A ZCMT2-020D-A ZCMTK/020C-AE ZCMT1-020D-A ZCMTK/020C-A ZCMT3-020D-AE ZCMT1-020E-AE ZCMT1-020C-AE ZCMT3-020E-A ZCMT2-020E-AE
76495-898EA 2104.68 USD InStock
76495-898 76495-876 76495-886 76495-896 76495-874 76495-884 76495-902 76495-912 76495-878 76495-900 76495-910 76495-888 76495-890 76495-872 76495-894 76495-882 76495-892 76495-870 76495-880 76495-906 76495-916 76495-904 76495-914 76495-908
TETPOR AIR Filter Cartridges, Parker Bioscience
Filters Filter Cartridges
TETPOR AIR sterilization filter cartridges offer exceptional filtration performance while providing the highest levels of biosecurity throughout the process industry.
  • Assured biosecurity with absolute rated filtration
  • High flow rates with low pressure drops
  • High voids volume PTFE membrane
  • Unique prefilter layer
Operating at ambient temperature conditions, TETPOR AIR filter cartridges provide a cost-effective filtration solution. A unique polypropylene prefilter layer extends service life in heavily contaminated environments.
TETPOR AIR filter cartridges also utilize a well-proven, inherently hydrophobic expanded PTFE membrane validated as sterilizing grade in liquid in accordance with ASTM F838-05. This ensures the removal of all airborne bacteria, viruses and bacteriophage.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty