Would you like to log in again?
CARTRIDGE 5IN 3UM PP BF/226 BAYONET EPDM
Supplier: PARKER HANNIFIN TE
Cartridge, Nominal length: 5in, 3um Pleated Polypropylene Depth Media, SOE BF/226 Bayonet end cap with EPDM o-rings, designed for a wide range of prefiltration duties within the production of pharmaceuticals.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
CARTRIDGE 5IN 3UM PP BF/226 BAYONET EPDM 76496-362 PCPPK-03C-PE Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty