Would you like to log in again?
CARTRIDGE 5IN 0.2UM PES FIN/222 SILCN
Supplier: PARKER HANNIFIN TE
Cartridge, Nominal length: 5in, 0.2um Polyethersulphone membrane, SOE Fin/222 end cap with external silicone o-rings, have been specifically designed for the fast and cost-effective bioburden reduction of pharmaceutical solutions.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
CARTRIDGE 5IN 0.2UM PES FIN/222 SILCN 76496-654 ZCBRK-020D-PS Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty