Would you like to log in again?
CARTRIDGE 10IN 1UM PP BF/226 BAYONET SIL
Supplier: PARKER HANNIFIN TE
Cartridge, Nominal length: 10in, 1.0um Pleated Polypropylene media, SOE BF/226 Bayonet end cap with external silicone o-rings, utilized for the clarification and prefiltration of a wide range of products in the fermentation and fine chemical industries.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
CARTRIDGE 10IN 1UM PP BF/226 BAYONET SIL 76496-750 ZCPP1-1.0C Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty