Would you like to log in again?
CARTRIDGE 20IN 10UM PP FLAT TOP/222 SIL
Supplier: PARKER HANNIFIN TE
Cartridge, Nominal length: 20in, 10um Pleated Polypropylene media, SOE Flat Top/222 end cap with external silicone o-rings, utilized for the clarification and prefiltration of a wide range of products in the fermentation and fine chemical industries.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
CARTRIDGE 20IN 10UM PP FLAT TOP/222 SIL 76496-996 ZCPP2-010E Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty