Would you like to log in again?
CARTRIDGE 10IN 0.2UM PTFE BF/226 SILCN
Supplier: PARKER HANNIFIN TE
Cartridge, Nominal length: 10in, .2um Pleated PTFE membrane, SOE BF/226 Bayonet end cap with external silicone o-rings, sterile gas filters combine proven depth filter technology and a pleated construction to provide retention down to 0.01 micron in gas.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
CARTRIDGE 10IN 0.2UM PTFE BF/226 SILCN 76497-120 ZCHB-1C Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty