Would you like to log in again?
MINI CAPSULE STANDARD SIZE 2.5UM PP CS4
Supplier: PARKER HANNIFIN TE
Mini Capsule, Standard Size, with 2.5um Pleated Polypropylene Depth Media, 1/4in NPT (Male) connections and vent/draing valves with silicone seal, encapsulated design maximizes efficiency by providing faster, easier change-out without laborious cleaning procedures.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
MINI CAPSULE STANDARD SIZE 2.5UM PP CS4 76497-432 22-CSBPP-025-2 Case of 4 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty