Would you like to log in again?
CARTRIDGE 30IN 0.1UM PES FIN/222 EPDM
Supplier: PARKER HANNIFIN TE
Cartridge, Nominal length: 30in, 0.1um Polyethersulphone membrane, Fin/222 end cap with external EPDM o-rings. PROPOR MR filters have been specifically designed for fast, effective and economical removal of mycoplasma from cell culture media in the biopharmaceutical industry.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
CARTRIDGE 30IN 0.1UM PES FIN/222 EPDM 76498-058 ZCMR3-610D-PE Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty