Would you like to log in again?
GRAB-EEZ® Cleanroom Wipe Refills
Supplier: High-Tech Conversions
GRAB-EEZ®
GE-NT1-99
76498-087CS 326.62 USD InStock
76498-087
GRAB-EEZ® Cleanroom Wipe Refills
Wipes Cleanroom Wipes Dry Cleanroom Wipes
GRAB-EEZ® cleanroom wipe refills are designed with simplicity being a top priority. Typically, cleanroom wipes are supplied in flat sheets and placed on a table or in trays within a cleanroom.
  • Designed to eliminate wasteful use
  • Unique double C-fold cleanroom wipe
  • Suitable for ISO 5 (Class 100) or higher
  • Refill packaging designed for use with the GRAB-EEZ® cleanroom wipe dispenser
Typically, cleanroom wipes are supplied in flat sheets and placed on a table or in trays within a cleanroom. The unique double C-folded GRAB-EEZ® cleanroom wipe refills are stacked inside the GRAB-EEZ® cleanroom wipe dispenser unit, protected from the environment until use.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty