Would you like to log in again?
Coplin Staining Jar, Mortech®
Supplier: Mortech
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
BA002 BA003
76498-152EA 36.7 USD InStock
76498-152 76498-154
Coplin Staining Jar, Mortech®
Jars Staining Jars
Heavy, clear glass jar with four molded internal grooves to hold up to ten slides.
  • Glass base construction
  • Four molded grooves in glass hold up to ten 3×1" slides
Jar features a broad base to prevent spillage. Both glass and plastic lid options protect against contaminants. The plastic option is a tight-fitting screw lid to prevent fluid evaporation. The glass cover option simply and easily sets upon the jar.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty