Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Stainless Steel Ruler, Mortech®

Supplier: Mortech

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

BA038 BA036 BA037
76498-214EA 86.23 USD InStock
76498-214 76498-216 76498-212
Stainless Steel Ruler, Mortech®
Rulers
Rulers have a non-glare satin finish with micro-fine graduations etched on the surface.

  • Dimensions:
  • 150 and 300 mm long: 13 mm (0.5") wide;
  • 450 mm long: 19 mm (0.75") wide
  • Hardened and tempered stainless steel construction

Graduations are provided in increments of 0.5 mm on the top edge and full millimeters on the bottom edge.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty