Would you like to log in again?
Arbor Wrench, Mortech®
Supplier: Mortech
BD107
76498-370EA 41.2 USD InStock
76498-370
Arbor Wrench, Mortech®
Wrenches
This wrench is to be used in removing and installing new blades on select Mopec autopsy saws.
  • 0.5" combination
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty