Would you like to log in again?
ClearSteel™ Stainless Steel Cleaning and Polishing Wipes, Mortech®
Supplier: Mortech
ClearSteel™
BE039
76498-456EA 79.83 USD InStock
76498-456
ClearSteel™ Stainless Steel Cleaning and Polishing Wipes, Mortech®
Wipes Disposable Wipes and Towels Presaturated Wipes
Bring out the natural luster in equipment while leaving a protective covering that repels water, fingerprints, and dust. 160 premoistened towelettes per pop-up canister.
Specifically designed to be used on all equipment such as workstations, cabinets, coolers, and storage areas. Simply pull a wipe from the canister and with moderate pressure, clean the soiled area and allow to dry for 5 minutes. Wipe in the direction of the stainless steel grain for best results.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty