Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Super-Duty Body Bag, with Handles, Mortech®

Supplier: Mortech
BE081
76498-502EA 152.93 USD InStock
76498-502
Super-Duty Body Bag, with Handles, Mortech®
Bags Emergency Bags
Super-duty body bag.

  • Overall dimensions are 36W×94"L
  • Blue inner liner enables forensic photos
  • Envelope-style closure with nylon rust-resistant zipper
  • Bags feature 6 strong, 2" wide web handles with zinc plated fastemers
  • Capacity is 450 lbs.
  • Suitable for cremation

The material construction of these super-duty bags features a 12 mil polyethylene coated outer shell (silver), coupled with a 10 mil polyethylene coated inner liner (blue). Both layers are tear- and puncture-resistant. The seams are folded, sewn, and sealed for protection and leakage. The bags comply with OSHA reg. 3130.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty