Would you like to log in again?
Formalin Neutralizer Pads, Mortech®
Supplier: Mortech
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
BE094 BE095 BE093
76498-516PK 349.33 USD InStock
76498-516 76498-512 76498-514
Formalin Neutralizer Pads, Mortech®
Spill Control Materials
Dramatically minimizes exposure to formaldehyde vapors.
  • Minimizes cleanup
  • Incineration of pads is clean and efficient with minimal threat of exposure
  • Fiber absorbs and then locks in liquids so that work areas stay drier
  • Resists tearing, even when saturated
  • Internal sliding shelf for easy positioning cube for pouring and storage
  • Simple and easy to use
A major crisis facing today's hospitals, laboratories and other healthcare providers is the occupational exposure and disposal of formaldehyde, a known carcinogen. Formaldehyde Neutralizer Pads offer a way to significantly reduce exposure by chemically neutralizing formaldehyde and encapsulating the escaping vapors with the pad.
Clinical Pad applications include surgical cutting board pads, dissecting pads, autopsy table pads, and liners for body trays and cadaver bags. Formaldehyde Neutralizer Pads reduce exposure by chemically bonding with formaldehyde, resulting in the encapsulation of vapors-ideal for grossing areas and autopsy work.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty