Would you like to log in again?
EZ-Zyme® Instrument Cleaner Packets, Mortech®
Supplier: Mortech
EZ-Zyme®
BE100
76498-518PK 250.12 USD InStock
76498-518
EZ-Zyme® Instrument Cleaner Packets, Mortech®
Cleaning Agents Detergents and Surface Cleansers
This product is one of the best enzyme cleaners on the market. Although not a disinfectant, it does dissolve all tissue debris, leaving every crack and crevice of dissection instruments looking brand new.
  • Contains 32 packets
Simply open the packet of concentrate, dilute into a gallon of warm tap water and place any hard-to-clean instrument into the solution. There is absolutely no hand scrubbing required.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty