Would you like to log in again?
Sterile Test Tubes, Caplugs
Supplier: Caplugs
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
222-2070-010 222-2061-050 222-2376-08I 222-2088-050 222-2095-05I 222-2380-080 222-2068-05I 222-2376-080 222-2094-050 222-2036-050 222-2003-010 222-2381-080 222-2012-010 222-2393-080 222-2394-08I 222-2068-050 222-2089-05I 222-2367-080 222-2099-050 222-2036-05I
76498-768CT 294.11 USD InStock
76498-768 76498-758 76498-780 76498-770 76498-792 76498-790 76498-766 76498-788 76498-778 76498-786 76498-764 76498-776 76498-762 76498-784 76498-774 76498-796 76498-760 76498-782 76498-772 76498-794
Sterile Test Tubes, Caplugs
Tubes Test Tubes
Sterile culture tubes are precision-molded from premium-grade, chemically inert plastic materials: polystyrene for clarity and polypropylene for toughness and chemical resistance.
  • Gamma radiation sterilized and shipped ready-to-use
  • Polystyrene tubes are certified to 8200×g
  • Round bottom
Polypropylene material is almost transparent and withstands centrifugation to 10000×g.
Ordering information: Caps are also sold separately; standard stopper caps and 2-position caps.
Packaging: All tubes are packaged in easy-to-open bags of 25 tubes each.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty