Would you like to log in again?
CO₂ Resistant Shakers, Thermo Scientific
Supplier: Thermo Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
88881103B 88881101 88881101B
76498-936EA 4220.31 USD InStock
76498-936 10753-628 76498-940
CO₂ Resistant Shakers, Thermo Scientific
Shakers and Mixers Orbital Shakers
Get reliable around-the-clock operation ideally suited to keep your cells alive and flourishing within your working environment with Thermo Scientific™ CO₂ Resistant Shakers.
  • Orbital shaking through magnetic power eliminates the need to replace belts, available in either a 3 or 19 mm orbit
  • Specially treated mechanical components protect up to 20% CO₂ and 95% humidity conditions
  • Convenient external control box with 4-20mA data output port provides added security and remote monitoring capability
  • Minimal heat dissipation assures conditions within the CO₂ chamber are not effected
  • Stainless steel construction for protection from corrosive CO₂ gassing conditions
  • CO2 resistant shaker can be used outside of CO₂ incubator for maximum value
  • External control box for ease-of-use without disrupting your samples or opening the door
Unlike a combined shaking incubator, a CO₂ resistant shaker can be easily removed from the incubator to facilitate cleaning and disinfection, protecting your samples.
With a 3 or 19 mm orbit size option, they have wide applications such as mammalian cell culture, bacterial suspensions, staining and destaining, washing procedures, hybridization, solubility studies and extraction procedures.
Accessories information: Universal platform (10753-670) accommodates a maximum 30×50, 15×100, 15×150, 15×200, 15×250, 9×500, 6×1000, 4×2000, 2×3000 or 1×5000 ml flasks, 6×500, 6×1000 infusion bottles, 2 funnel clamps, 6×96 well microplate holders, 1× spring wire rack, 4× sticky mats or 2 fixed/adjustable test tube racks.
Universal platform (76498-944) accommodates a maximum 49×50, 25×100, 25×150, 18×200, 18×250, 10×500, 7×1000, 4×2000, 4×3000 or 2×5000 ml flasks, 12×500, 9×1000 infusion bottles, 6 funnel clamps, 10×96 well microplate holders, 1× spring wire rack, 9× sticky mats or 4 fixed/adjustable test tube racks.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty