Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

CO₂ Resistant Shakers, 220 to 240 V

Supplier: Thermo Scientific

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

88881102B 88881102 88881104B
10753-630EA 6439.31 USD InStock
10753-630 76498-938 76498-942
CO₂ Resistant Shakers, 220 to 240 V
Shakers and Mixers Orbital Shakers
Get reliable around-the-clock operation ideally suited to keep your cells alive and flourishing within your working environment with Thermo Scientific™ CO₂ Resistant Shakers.

  • 220 to 240 V model
  • Orbital shaking through magnetic power eliminates the need to replace belts, available in either a 3 or 19 mm orbit
  • Specially treated mechanical components protect up to 20% CO₂ and 95% humidity conditions
  • Convenient external control box with 4 to 20 mA data output port provides added security and remote monitoring capability
  • Minimal heat dissipation assures conditions within the CO₂ chamber are not effected
  • Stainless steel construction for protection from corrosive CO₂ gassing conditions
  • CO2 resistant shaker can be used outside of CO₂ incubator for maximum value
  • External control box for ease-of-use without disrupting your samples or opening the door

Unlike a combined shaking incubator, a CO₂ resistant shaker can be easily removed from the incubator to facilitate cleaning and disinfection, protecting your samples.

With a 3 or 19 mm orbit size option, they have wide applications such as mammalian cell culture, bacterial suspensions, staining and destaining, washing procedures, hybridization, solubility studies and extraction procedures.

Accessories information: Universal platform (10753-670) accommodates a maximum 30×50, 15×100, 15×150, 15×200, 15×250, 9×500, 6×1000, 4×2000, 2×3000 or 1×5000 ml flasks, 6×500, 6×1000 infusion bottles, 2 funnel clamps, 6×96 well microplate holders, 1× spring wire rack, 4× sticky mats or 2 fixed/adjustable test tube racks.
Universal platform (76498-944) accommodates a maximum 49×50, 25×100, 25×150, 18×200, 18×250, 10×500, 7×1000, 4×2000, 4×3000 or 2×5000 ml flasks, 12×500, 9×1000 infusion bottles, 6 funnel clamps, 10×96 well microplate holders, 1× spring wire rack, 9× sticky mats or 4 fixed/adjustable test tube racks.

Caution: 220 to 240 V model. Corresponding model with voltage suitable for use in US (10753-628, 76498-940 and 76498-942) is also available, please see separate product page.

Order Now
 

ORDER

SPECIFICATIONS

ACCESSORIES

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty