Would you like to log in again?
GLOVES HYFLEX 11-290 SZ18 NARROW PR CS30
Supplier: Ansell Healthcare
Gloves, Black Intercept (Kevlar, stainless steel, polyamide, spandex ), 15G Circular Knit (Patented Technology), Elasticized Top, Knitwrist Cuff, Narrow Width, Integrated elbow pad to protect against impact, Fiber glass free, Size: 18in
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
GLOVES HYFLEX 11-290 SZ18 NARROW PR CS30 76498-956 11290180-N Case of 30 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty