Would you like to log in again?
GLOVES HYFLEX 11-757 SIZE 10 PR PK12
Supplier: Ansell Healthcare
Gloves, 18 Gauge (INTERCEPT) Nylon, Stainless Steel, Tungsten, Polyester Seamless Knit Liner, Polyurethane Palm Coat, Knitwrist Cuff, touchscreen compatible to ensure that workers do not need to change or remove gloves between tasks, Size 10
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
GLOVES HYFLEX 11-757 SIZE 10 PR PK12 76498-968 11757100 Case of 1728 Retrieving
Pack of 12 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty