Would you like to log in again?
O DIN S-1
Supplier: IKA WORKS
DIN spindle set (O-DINS-1) measures viscosities according to DIN 53019 and can be used with customer-specific vessels, includes a single-wall-open-jacket to quickly dive into sample liquid as well as coaxial cylinder system for analysis of sample, maximum measurable viscosity is 50,000 mPas.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
O DIN S-1 76499-048 25000326 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty