Would you like to log in again?
ELVAS-1
Supplier: IKA WORKS
Elvas-1, extremely low viscosity adapter spindle set includes a double jacket to quickly heat up or cool down the sample as well as a coaxial cylinder system for the analysis of the sample at a specific shear rate and shear stress, viscosity is 2000 mPas.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
ELVAS-1 76499-052 25000390 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty