Would you like to log in again?
SP SET-2 STANDARD SPINDLE
Supplier: IKA WORKS
Standard spindle set SP set-2, includes six spindles (SP7, SP8, SP9, SP10, SP 11, SP12). It is particularly suitable for viscosity determination in combination with the rotary viscometers ROTAVISC me-vi, hi-vi I or hi-vi II Complete or rather ROTAVISC me-vi, hi-vi I or hi-vi II Advanced.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
SP SET-2 STANDARD SPINDLE 76499-062 25000329 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty