Would you like to log in again?
VOL-SP-2.1 SPINDLE
Supplier: IKA WORKS
VOL-SP-2.1 is an optional spindle for an extended viscosity range. It is especially suitable for sample volumes of 2.1 ml, It is required to use the 2.1 ml chamber VOL-C-RTD-2.1 in combination with the VOL-SP-2.1 spindle.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
VOL-SP-2.1 SPINDLE 76499-082 25007061 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty