Would you like to log in again?
ROTAVISC Oils for Rotational Viscometers, IKA
Supplier: IKA
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
ROTAVISC
25000399 25000469 25000435 25000468 25000436 25000397 25000466 25000437 25000467 25000434 25000398
76499-098EA 154.99 USD InStock
76499-098 76499-118 76499-108 76499-114 76499-104 76499-116 76499-106 76499-110 76499-100 76499-112 76499-102
ROTAVISC Oils for Rotational Viscometers, IKA
Lubricants
These certified standard silicone oils can be used to check the measurement accuracy of ROTAVISC.
Delivery information: Supplied with certificate.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty