Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

PROSAT® Sterile Polywipe-C Heatseal Wipes, Contec®

Supplier: Contec

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

PROSAT®
PSCS0009SZ PSCS0009 PSCS0012
76422-126CS 864.91 USD InStock
76422-126 76463-334 76499-222
PROSAT® Sterile Polywipe-C Heatseal Wipes, Contec®
Wipes Cleanroom Wipes Presaturated Cleanroom Wipes
100% interlock knit standard weight textured polyester wipes presaturated with 70% USP grade IPA and 30% DI water.

  • Recommended for Grade A/B or ISO Class 3-8 environments
  • Standard weight laundered knitted 100% polyester fabric for extremely low levels of particles and fibers
  • Heatsealed edges prevent fiber generation during use
  • Small number of wipes per resealable pouch to eliminate waste
  • Presaturated wipe reduces VOC emissions and solvent use
  • Resealable pouch preserves cleanliness and solvent saturation levels

PROSAT® Sterile™ Polywipe-C Heatseal Wipes are made from 100% knitted polyester that is laser cut to bond the fibers at the edges of the wipe. The wipe is chemical-resistant and exceptionally low in particles and extractable residue, making it ideal for critical cleaning.

These wipes are presaturated with 70% USP grade isopropanol and 30% DI water, which greatly reduces solvent usage and reduces the VOCs associated with spraying IPA. Using presaturated wipes also maximizes process control and improves consistency between operators. The wipes are gamma irradiated and validated per AAMI Guidelines to a 10⁻⁶ SAL, allowing use in the highest grade life science cleanrooms.

The wipes are provided in convenient and easy to use peel and reseal pouches. The resealable pouch with the option of a small number of wipes per pouch helps reduce waste and control saturation volume and cleanliness.

Packaging: Wipes are packaged 20 per pouch, 24 pouches per case.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty