Would you like to log in again?
FROCK CLEANROOM BULK N-STERILE XL CS50
Supplier: CHEMLAND-LAKELAND INDUS
Frock, Cleanroom, Bulk, Nonsterile, Zipper Front, Elastic Wrist With Thumb Loop, 99.999% Bacterial and Particle Filtration Efficiency, Suitable for ISO CLass 4 and higher applications, USP 795 and USP 797 Compliant, Sterility Assurance Level of 10-6, Size: XL
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
FROCK CLEANROOM BULK N-STERILE XL CS50 76499-232 CTL191CMB-XL Case of 50 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty