Would you like to log in again?
CE Duo® CleanTech® Sterile 6-Pack with Bucket Liner, Vileda Professional®
Supplier: Vileda Professional
169581
76499-312CS 1784.69 USD InStock
76499-312
CE Duo® CleanTech® Sterile 6-Pack with Bucket Liner, Vileda Professional®
Mops
Mop is available in a conveinent six pack with a bucket liner, cutting down not only on the time it takes to prepare the mops but also significantly reducing the amount of packaging waste.
  • Microfiber, overstitched cleanroom foam for cleaning/disinfecting floors, walls, and ceilings
  • Adhesive free lamination techniqies are used to prevent additional contamination
  • Excellent microfiber cleaning results have been independently proven
  • Validated to 10⁻⁶ SAL per ISO 11137
  • Reduce packaging waste
  • Reduce amount of time needed to pre-prepare the mops
The Sterile CE Duo® CleanTech® mop is a single-use, double-sided microfiber cleanroom foam mop for cleaning and disinfecting floors, walls and ceilings. Now available in a conveinent 6 pack with included bucket liner.
Packaging: Packaged 6 sterile mops per pack, double bagged with bucket liner.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty