Would you like to log in again?
Replacement Shelf and Polyethylene Trays/Sumps for Justrite® Safety Cabinets, Justrite®
Supplier: Justrite
29050 29057 29059 29053 29058 29061 29063 29054 29060 29052 29055 29051 29062
76499-360EA 239.29 USD InStock
76499-360 76499-350 76499-362 76499-356 76499-368 76499-346 76499-358 76499-348 76499-352 76499-364 76499-354 76499-344 76499-366
Replacement Shelf and Polyethylene Trays/Sumps for Justrite® Safety Cabinets, Justrite®
Furniture Shelves
Polyethylene trays over shelves add corrosion protection when storing corrosive liquids.
  • Resistant to aggressive chemicals
  • Polyethylene trays sit on top of galvanized steel shelves
  • Slightly curved edge provides additional protection
  • Patented SpillSlope® galvanized steel shelves direct spills to back and bottom of a leakproof sump
Justrite galvanized steel shelves hold up to 159 kg (350 lbs.) and adjust on 76 mm (3") centers to meet specific workflow needs. Customize your cabinet by adding shelves to increase storage space and accommodate different sized containers. Place polyethylene trays over shelves for corrosion protection when storing corrosive liquids.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty