Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Duo-i Dual Systems, Ultrapure (18.2 MΩ-cm) and DI Lab Water, Avidity Science

Supplier: Avidity Sciences

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

7120-4500-443 7120-4500-433 7120-4500-493 7120-4500-443 7120-4500-473
76499-370EA 17683.37 USD InStock
76499-370 76499-372 76533-356 76509-502 76509-500
Duo-i Dual Systems, Ultrapure (18.2 MΩ-cm) and DI Lab Water, Avidity Science
Water Purification Systems Pure and Ultra Pure Water Systems
Two-In-One system provides DI or RO water to a storage tank, and capable of dispensing Type 1 and Type 2 or Type 3 water from dispense guns (depending on model variation).

  • Fast, volumetric and variable dispense up to 2 L/min
  • Ability to dispense Type 1 and Type 2 water (optional) from remote dispenser(s)
  • Make-up flow rate of 10 L/h
  • Unique twin pass RO technology ensures enhanced DI cartridge capacity
  • Data capture with printable option
  • Integrated water leakage protection
  • Easy to change consumables with traceability and data tags
  • Interactive 7" touch screen
  • Color coded and adjustable dispensing stations with up to two remote dispensing options
  • Choice of storage reservoir size - 35, 70, or 105 L

The innovative and unique Duo-i provides two water qualities within one easy to use system. Save on space, initial outlay and consumable costs. Type 2 or 3 water (based on model) is available from the reservoir while Type 1 water is ready to dispense from the remote dispenser. Additional dispense guns allow dispensing of water from reservoir. This model filters water into a tank at 10, 20, or 30 L per hour (based on model) which can be dispensed from a 35, 70, or 105 L reservoir.

Application: Duo-i II.I is suitable for Type 2 water usage. Type 2 water is used for buffer and media production, sample and reagent diluent, general chemistry, protein electrophoresis, histology, cytology, and spectrophotometry. Duo-i III.I Type 3 water is suitable to feed an ultrapure water machine, glass washer, humidity/stability chambers and other laboratory equipment. Duo-i II.I and III.I is also suitable for Type 1 water usage. Type 1 water is used for molecular biology, electrophoresis, cell and tissue culture, sequencing, HPLC, genomics, pharmacology, and any other sensitive applications.

Certifications: Manufactured to ISO 9001:2015 standard. Certified to cUL, and IEC 61010-1:2010+A1.

Ordering information: Endotoxin and nuclease-free point of use filter available (see 76337-284). First round of consumables are included with purchase of unit. Ships within 2 to 3 weeks and installed within 4 to 6 weeks from receiving the PO.

Order Now
 

ORDER

SPECIFICATIONS

ACCESSORIES

SERVICES

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty