Would you like to log in again?
CENTRIFUGE BRUSHLES 6X50 OR 10/15ML 110V
Supplier: C & A SCIENTIFIC CO INC
Centrifuge, Brushless, 6 X 50ml, Includes Adapters For 10/15ml, Max Speed 4,000 rpm, 110V, 60Hz
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
CENTRIFUGE BRUSHLES 6X50 OR 10/15ML 110V 76499-388 XC-2500VM Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty