Would you like to log in again?
EMS Style 277 Tweezers, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
2ABCPR.SA 7CPR.SA 259CPR.SA 2ACPR.SA 5CPR.SA 2ABCFR.SA 4WFCPR.SA 5XCPR.SA
76499-504EA 45.32 USD InStock
76499-504 76499-506 76499-528 76499-502 76499-546 76499-672 76499-496 76499-540
EMS Style 277 Tweezers, Electron Microscopy Sciences
Forceps
Tweezer with durable and self-aligning replacement tip, and anti-magnetic stainless steel handle.
  • Specially designed for the electronics industry
  • Excellent for handling sensitive electronic components
  • Ideal for ceramic and glass parts where non-metallic non-scratching is critical
Carbon Fiber tips are intended for handling sensitive components and devices (electronic components, micro-mechanical parts, glass and ceramic substrates, capillary, etc.). They offer high rigdity, self lubricating properties, excellent wear, abrasion resistance, and are ESD safe. They also provide excellent chemical resistance to most aggressive substances. Not resistant to strong acids, alkalis, and hot water or steam.
PEEK tips are reinforced with carbon fiber, and are intended for handling components in cleaning/chemical/assembly processes. They may also be used at high temperature (soldering). PEEK tips have excellent resistance to chemicals and aggressive agents, as well as excellent resistance to thermal aging. They are ESD safe.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty