Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
EMS Zirconia Ceramic Tip Tweezers, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
78127-73 78127-2A 78127-74-T 78127-2A-T 78127-71 78127-7 78127-73-T 78127-72-T 78127-72 2AZJK.SA 78127-74 78127-71-T 78127-7-T 72ZJ.SA
76376-124EA 63.08 USD InStock
76376-124 76499-592 76376-114 76376-116 76376-118 76376-120 76376-122 100494-610 76499-508 100494-608 100494-606 100494-614 100494-604 100494-612
EMS Zirconia Ceramic Tip Tweezers, Electron Microscopy Sciences
Forceps
These tweezers have Zirconia ceramic precision tips.
  • Light weight
  • Aluminum handles
Surface resistively 10E12 ohm, Contamination free, high temperature (1000 °C). All styles have length of 134 mm (5¹/₄").
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty