Would you like to log in again?
Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Saturday, May 25. 
Bio-Paper Folding Biopsy Papers, Trifold and Ova, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
62326-36 62326-35 62326-37
76499-580PK 122.77 USD InStock
76499-580 76499-582 76499-578
Bio-Paper Folding Biopsy Papers, Trifold and Ova, Electron Microscopy Sciences
Biopsy Pads
These perforated filtered Biopsy Papers secures biopsies during processing.
Tri-Fold Biopsy Paper folds once and then again, allowing it to fit snugly and securely into cassettes.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty