Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.

PEN Foil Bottom Sterile Dishes, Electron Microscopy Sciences

Supplier: Electron Microscopy Sciences
70665-24
76499-586PK 389.34 USD InStock
76499-586
PEN Foil Bottom Sterile Dishes, Electron Microscopy Sciences
Trays/Dishes/Pots Laboratory Trays/Dishes
PEN foil bottom sterile dishes are ideal for laser microdissection.

  • PEN foil bottoms
  • Glass well diameter: 30 mm
  • Diameter: 50 mm
  • Height: 7 mm

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty