Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Stainless Steel Probes, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
78325-91 78326-10 78325-93 78326-06 78326-09 78326-07 78326-11 78326-08
76499-628EA 6.93 USD InStock
76499-628 76499-624 76499-636 76499-638 76499-620 76499-632 76499-634 76499-630
Stainless Steel Probes, Electron Microscopy Sciences
Dissection Needles
Stainless steel probes are perfect for lab applications and electronics inspections.
  • 155 mm overall length
  • Multiple tip styles available
Stainless steel probes are made of rust-free stainless steel and have a serrated handle for secure grip.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty