Would you like to log in again?
Au-Flat Grids, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
Au-Flat
AUFT222-50 AUFT313-50 AUFT313-05
76499-690PK 1581.04 USD InStock
76499-690 76499-686 76499-688
Au-Flat Grids, Electron Microscopy Sciences
Graticules Reticles and Grids
Au-Flat, ultrastable Cryo-EM sample support with a 45 nm holey Gold alloy film on 3 mm gold mesh grid.
  • Holey Au/Pd film on a gold mesh grid
  • Stronger than carbon films
  • More capable of surviving the Cryo-EM workflow
The Au-Flat Cryo-EM grid significantly reduces beam-induced motion during imaging compared to carbon films, improving both image quality and resolution.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty