Would you like to log in again?
Epoxy Coated Tweezers, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
5.SA.NE 2A.SA.NE 3C.SA.NE K3.SA.NE 00.SA.NE K5F.SA.NE SM103.SA.NE 15A.C.NE 1.SA.NE 15ARW.C.NE SM108.SA.NE SM104.SA.NE 4.SA.NE 15AGW.C.NE 3.SA.NE K4F.SA.NE 54MW.C.N K6F.SA.NE 15AGHM.C.NE 14AGW.C.NE
76499-492EA 112.05 USD InStock
76499-492 76499-482 76499-450 76499-490 76499-774 76499-488 76499-500 76499-874 76499-522 76499-534 76499-770 76499-484 76499-870 76499-772 76499-486 76499-872 76499-518 76499-536 76499-768 76499-516
Epoxy Coated Tweezers, Electron Microscopy Sciences
Forceps
Epoxy coated tweezers have an impact-resistant surface with excellent flow and good elasticity.
  • Good resistance to many diluted acids and alkalis
  • Can operate continuously at temperatures up to 120 °C
Compromised of polyester and epoxy resins, plus the electrically conductive additives.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty