Would you like to log in again?
NIGHTSEA Retrofit Lamp Base Controls, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
SFA-BASE-PULSE SFA-BASE-DIM-R
76499-826EA 580.67 USD InStock
76499-826 76499-828
NIGHTSEA Retrofit Lamp Base Controls, Electron Microscopy Sciences
Lights
NIGHTSEA lamp base control options.
NIGHTSEA DIM option lamp bases feature an OFF/ON/DIM switch to change the intensity/power of the light. This allows a finer control in the brighter portion of the adjustment range. Intensity indicator from 1 to 10.
NIGHTSEA PULSE option adds a BNC connector on the rear of the base that accepts a user-supplied voltage input to turn the light on and off. Voltage input is 2.8 to 6.0 VDC for 'ON', <0.6 VDC for 'OFF'. Maximum operation frequency of 10 kHz.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty