Would you like to log in again?
SP-1 STANDARD SPINDLE
Supplier: IKA WORKS
SP-1 Standard spindle, Cylinder, Thread M2.5 LH, Diameter: 18.8 mm, Immersion depth: 81 mm, Spindle length: 115 mm, Dimensions: (W x H x D)20 x 117 x 20 mm
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
SP-1 STANDARD SPINDLE 76499-898 0025006828 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty