Would you like to log in again?
BATH IB R 9 ECO PLASTIC RECTANGULAR 9L
Supplier: IKA WORKS
Bath, IB R 9 eco, Plastic, rectangular, size S, 9 liter volume, Max. Temperature: 100 deg C; water only, Inner dimensions: 307 x 239 x 151 mm, Outer dimensions: 356 x 287 x 167mm
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
BATH IB R 9 ECO PLASTIC RECTANGULAR 9L 76499-900 0020004382 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty