Would you like to log in again?
ROTAVISC HI-VI I HELI COMPLETE
Supplier: IKA WORKS
ROTAVISC hi-vi I HELI Complete, determines the viscosity of highly viscous liquids in all areas of application ranging from the laboratory to quality control, includes a standard spindle set (SP 7-SP 12), a protective bracket, temperature sensor and HELISTAND stand incl. T-spindle set.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
ROTAVISC HI-VI I HELI COMPLETE 76499-922 0025008217 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty