Would you like to log in again?
Clorox® Healthcare Bleach Germicidal Wipes, Essendant
Supplier: Essendant
Clorox®
CLO30577CT
76500-082CT 126.9 USD InStock
76500-082
Clorox® Healthcare Bleach Germicidal Wipes, Essendant
Wipes Disposable Wipes and Towels Disinfectant Wipes
Healthcare professionals trust Clorox® Healthcare Bleach Germicidal Wipes to help overcome infection control challenges in a constantly changing healthcare environment. Clorox® Healthcare Bleach Germicidal clinical wipes are a ready to use, one step cleaner containing a 1:10 bleach dilution to meet CDC and OSHA guidelines for disinfecting surfaces in healthcare settings.
  • Ready to use cleaner disinfectant; EPA registered to kill C. Difficile and 58 other microorganisms
  • Powerful disinfectant wipes are great for terminal cleaning
  • Use in healthcare settings including hospitals, medical facilities, dental offices, veterinary offices, and long-term care facilities
  • Meets CDC and OSHA recommendations
  • Tested for surface safety on common healthcare and office surfaces and equipment
Designed for daily disinfection, high-touch surfaces and medical equipment. These wipes are EPA registered to kill a total of 58 microorganisms, including HAI causing bacteria like MRSA and VRE in 30 seconds, outbreak causing viruses such as Influenza A and B viruses, Measles virus and Norovirus in 1 minute and C. difficile spores, TB and fungi in 3 minutes. These wipes kill 99.9999% of C. difficile spores, according to the current EPA recommended testing standard.
Wipes are engineered to minimize residue and leave less visible residue on surfaces. They are formulated with an anti-corrosion agent and are compatible with common healthcare surfaces including stainless steel, glass, plastic, porcelain, glazed ceramic, laminated surfaces, and vinyl. Intended for use in healthcare settings.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty