Would you like to log in again?
LYSOL® Clean and Fresh Multi-Surface Cleaner, Sparkling Lemon and Sunflower Essence, Essendant
Supplier: Essendant
LYSOL®
RAC78626CT
76500-100CT 55.53 USD InStock
76500-100
LYSOL® Clean and Fresh Multi-Surface Cleaner, Sparkling Lemon and Sunflower Essence, Essendant
Cleaning Agents Disinfectants
Approved to kill germs and odor-causing bacteria even when diluted. Kills 99.9% of viruses and bacteria on hard, non-porous surfaces when used as directed.
  • Cuts through tough grease and grime
  • Leaves a fresh clean scent
  • Chemical compound: n-Alkyl (C14 50%, C12 40%, C16 10%) Dimethyl Benzyl Ammonium Chloride
Use on floors, countertops, washable walls, door knobs, showers, bathtubs, diaper pails, toilet bowl exteriors, and appliance exteriors. This disinfectant cleaner works against bacteria, dust, germs, grease, grime, oil-based soils, organic matter, smoke film, soap scum, and water-based soils.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty