Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
ProEASYOFF Heavy-Duty Cleaner Degreaser Concentrate, Essendant
Supplier: Essendant
ProEASYOFF
RAC89771CT
76500-108CT 122.57 USD InStock
76500-108
ProEASYOFF Heavy-Duty Cleaner Degreaser Concentrate, Essendant
Cleaning Agents Detergents and Surface Cleansers
Heavy-duty cleanser is specially formulated to cut through tough kitchen grease, fat, and oil.
  • Easily removes tough oily soil from a wide variety of surfaces
  • Economical formula makes up to 128 gallons (1:128)
  • Thorough cleanser with a variety of applications
  • Chemical compound: C9-11 Alcohols Ethoxylate; Sodium Carbonate
This formula easily removes tough oily soil from a wide variety of surfaces and may be used in pressure washers and automatic scrubbers. Suitable for use on counters, tables, grinders, refrigerators, concrete and ceramic floors, hoods and grills, engines, machinery, and tools.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty