Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Diversey™ Easy Paks Neutral Cleaner, Essendant
Supplier: Essendant
Easy Paks®
DVO990653
76500-128CT 139.66 USD InStock
76500-128
Diversey™ Easy Paks Neutral Cleaner, Essendant
Cleaning Agents Detergents and Surface Cleansers
Bring back the shine to high-gloss floors without having to worry about harming them in the process.
  • Cleans and protects high-gloss finishes
  • Leaves no dulling film, even without rinsing
  • Pre-measured packet dissolves in water
  • Formula leaves a fresh pine forest scent
  • Packets supplied in a large, easy-to-store tub
Formula loosens and removes soil found in high-traffic areas, while protecting high-gloss, high solids finishes. Plus, the formula is phosphate-free and leaves no dulling film, without the need for rinsing. The premeasured packs of cleaning solution quickly dissolve in water, allowing users to start cleaning immediately. Ideal for cleaning surfaces ranging from terrazzo, marble, slate, vinyl and vinyl asbestos, asphalt, ceramic, and quarry tile. 90 Packets per tub.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty