Would you like to log in again?
Fabuloso® All-Purpose Cleaner, Lavender Scent, Essendant
Supplier: Essendant
Fabuloso®
CPC05253
76500-132CT 96.13 USD InStock
76500-132
Fabuloso® All-Purpose Cleaner, Lavender Scent, Essendant
Cleaning Agents Detergents and Surface Cleansers
Fabuloso Professional is an all-purpose cleaner and degreaser that is effective for a wide variety of cleaning applications.
  • 50% more cleaning power comparing active ingredients per drop vs. Fabuloso
  • Lavender scent
  • Deodorizes and degreases
  • Makes 64 gallons of ready-to-use solution
  • For use on multiple surface types
  • neutral pH
The Fabuloso Professional formula deep cleans to remove grease and soils while leaving a long-lasting fragrance. A neutral pH makes the product safe for surfaces. No rinsing step needed in floor cleaning applications.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty