Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Drano® Max Gel Clog Remover, Essendant
Supplier: Essendant
Drano®
SJN694768
76500-158CT 128.39 USD InStock
76500-158
Drano® Max Gel Clog Remover, Essendant
Cleaning Agents Detergents and Surface Cleansers
Thick bleach formula cuts right through standing water, clinging to the toughest clogs to blast them fast.
  • Pours through standing water straight to the clog
  • Works in 15-30 minutes
  • Includes an ingredient to protect pipes from corrosion
  • Can remain in drains overnight
Drano Max Gel Clog Remover works great to remove hair, soap scum, and other gunky clogs. Pours straight through standing water and works quickly to restore drains. It can be applied to metal, plastic, or PVC to remove food particles, grease, hair, oil, or organic matter. Unscented.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty