Would you like to log in again?
Clorox® Disinfecting Wipes, Crisp Lemon, Essendant
Supplier: Essendant
Clorox®
CLO01628 CLO01594CT
76500-450CT 97.52 USD InStock
76500-450 76500-196
Clorox® Disinfecting Wipes, Crisp Lemon, Essendant
Wipes Disposable Wipes and Towels Disinfectant Wipes
Disinfecting Wipes clean and disinfect in one easy step. These disinfecting wipes are EPA-registered to kill 99.9% of germs on hard nonporous surfaces, including cold and flu viruses, the COVID-19 virus (SARS-CoV-2), E. coli, Salmonella, Staph, Strep, MRSA, and the organism that can cause whooping cough.
  • Disinfecting wipes are triple-layered to clean, disinfect, and remove allergens
  • All-purpose disposable wipes remove common allergens, germs, and messes
  • Safe for finished wood, sealed granite, and stainless steel and on non-food-contact surfaces
  • Disinfect and deodorize with an all-in-one cleaning alternative that’s safe to use on toys and more
These wipes also remove common allergens like pet dander, pollen particles, and grass. One wipe kills most viruses in as little as 15 seconds on a wide variety of surfaces. The premoistened, thick, antibacterial wipes are perfect for spills and dirt on a variety of hard, nonporous surfaces including acrylic, glass, and vinyl. They quickly clean and disinfect high-touch germ hot spots such as doorknobs, handrails, and faucets, and are suitable for use on phones and keyboards. These sanitizing wipes are ideal for use in shared public spaces, offices, athletic facilities, schools, day care centers, busy healthcare environments, long term care, and other commercial facilities.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty